Learn More
Cayman Chemical SPE CartrIdgesC181 ml 5ea
SDP
Catalog No. 502942049
Encompass_Preferred
This product may be used for purification of various lipid mediators such as prostaglandins and hydroxylated fatty acids from various sample types prior to analysis by LC-MS/MS or ELISA. References are provided below for applicable C18 sample purification methods prior to LC-MS analysis.{19261,26615,8825,15162} For sample purification prior to analysis by ELISA, we recommend consulting the appropriate assay kit booklet for instructions.
Catalog No. 50-294-2049 Supplier Cayman Chemical Supplier No. 4000775 EA
This product may be used for purification of various lipid mediators such as prostaglandins and hydroxylated fatty acids from various sample types prior to analysis by LC-MS/MS or ELISA. References are provided below for applicable C18 sample purification methods prior to LC-MS analysis.{19261,26615,8825,15162} For sample purification prior to analysis by ELISA, we recommend consulting the appropriate assay kit booklet for instructions.