Learn More
Cayman Chemical SPE CartrIdgesMIxed Bed 10ea
SDP
Catalog No. 502942118
Encompass_Preferred
Mixed-mode cartridges packed with non-polar and strong anion exchange chemistries
Catalog No. 50-294-2118 Supplier Cayman Chemical Supplier No. 40002110 EA
Mixed-mode cartridges packed with non-polar and strong anion exchange chemistries