Learn More
Cayman Chemical SPE Cartridges (Mixed Bed), 25 ea
SDP
Catalog No. 502942000
Encompass_Preferred
Mixed-mode cartridges packed with non-polar and strong anion exchange chemistries
Catalog No. 50-294-2000 Supplier Cayman Chemical Supplier No. 40002125 EA
Mixed-mode cartridges packed with non-polar and strong anion exchange chemistries