Learn More
Cayman Chemical SPE CartrIdgesPhenyl 25ea
SDP
Catalog No. 502942067
Encompass_Preferred
Phenyl cartridges recommended for use with Angiotensin II EIA Kit.
Catalog No. 50-294-2067 Supplier Cayman Chemical Supplier No. 40004825 EA
Phenyl cartridges recommended for use with Angiotensin II EIA Kit.