Learn More
Solstice SPE Columns, Octyl (C8), End-capped, 900 mg
Catalog No. 6002893 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25
Catalog No. Quantity
600-28-93 25
1 options
Catalog No. 600-28-93 Supplier Solstice Supplier No. 9214
Add to Cart
Add to Cart
Medical-grade polypropylene with PE frits
  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25