Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPE Columns, THC (C8 + Aminopropyl), Medical-grade polypropylene with PE frits, 130 mg/1 mL, Solstice

Catalog No. 6002977 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 Ea.
Packaging:
FIBERBOARD BOX
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600-29-77 100 Ea. FIBERBOARD BOX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 600-29-77 Supplier Solstice Supplier No. 9910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

THC (C8 + Aminopropyl), Medical-grade polypropylene with PE frits, 130 mg/1 mL

  • Wide selection of support materials and multiple cartridge/column sizes
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SPE Columns
Name Note THC (C8 + Aminopropyl), Medical-grade polypropylene with PE frits, 130 mg/1 mL
CAS 1343-88-0, 7631-86-9, 63231-67-4, 68607-86-3, 7732-18-5
CAS Max % 100.0000%, 95.0000%, 100.0000%, 100.0000%, 5.0000%
Quantity 100 Ea.
Molecular Formula MgO3Si
Linear Formula NH4ClO4
MDL Number MFCD00011421
InChI Key ZADYMNAVLSWLEQ-UHFFFAOYSA-N
Molecular Weight (g/mol) 100.39
Grade Analytical
Packaging FIBERBOARD BOX
Density 1.95 g/mL at 25 °C(lit.)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.