Learn More
Hach Company ValuSep SPE Filter, 90 mm, 50/PK
Supplier:  Hach Company 2947802
Encompass
ValuSep™ SPE Filters for EPA Method 1664A are designed to provide high recovery and low blank values in Horizon SPEDEX and SPE Manifold applications. Chemically treated with a proprietary reverse phase adsorbent, single layer borosilicate glass ValuSep™ SPE Filters (47mm) can filter the 40 mg/L EPA PAR standard in approximately 10 minutes with significantly faster filtration timesfor 1 L NPDES-type samples. ValuSep™ SPE Filters offer high recoveries and low blanks at a great price.
Catalog No. 50-199-6936