Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Supelco™ SPE Frit Insertion Tool

Catalog No. 111011733 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Description:
For use with 3 mL SPE tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
11-101-1733 For use with 3 mL SPE tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-1733 Supplier MilliporeSigma™ Supelco™ Supplier No. 55218U

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Used to tighten or insert frits into SPE tubes.

TRUSTED_SUSTAINABILITY

Specifications

Description For use with 3 mL SPE tubes
For Use With (Application) Food and Beverages
Type SPE Accessory
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.