Learn More
Trajan Scientific SPE MEPS Kit BIN 5/PK
SDP
Catalog No. 5000494197
Encompass_Preferred
GC MEPS BIN development kit (1x each C18 C8 C2 Si C8 SCX) for CTC Analytics 250 L syringes 0 63 mm OD cone tipped needle PK5
Catalog No. 50-004-94197 Supplier Trajan Scientific Supplier No. 2900305
GC MEPS BIN development kit (1x each C18 C8 C2 Si C8 SCX) for CTC Analytics 250 L syringes 0 63 mm OD cone tipped needle PK5