Learn More
MilliporeSigma™ Supelco™ SPE Vacuum Pump Trap Kit
Catalog No. 111020836 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Visiprep SPE Vacuum Manifold
Catalog No. For Use With (Equipment)
11-102-0836 Visiprep SPE Vacuum Manifold
1 options
Catalog No. 11-102-0836 Supplier MilliporeSigma™ Supelco™ Supplier No. 57120U
Add to Cart
Add to Cart
For use with Visiprep SPE vacuum manifold
When installed between a Visiprep SPE vacuum manifold and the vacuum source, a Supelco SPE Vacuum Pump Trap collects all liquids that are aspirated through the SPE tubes, preventing contamination of the vacuum pump. The easily assembled kit contains a polypropylene filtering flask, a one-hole rubber stopper, 4 in. (10 cm) of polypropylene tubing and 5 ft. (1.5 m) of red rubber vacuum hose. The volume capacity of the Trap is approximately 1 L.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Food and Beverages
For Use With (Equipment) Visiprep SPE Vacuum Manifold
Type SPE Accessory
Volume (Metric) 1 L