Learn More
Speakman™ Eyesaver™ Wall-Mounted Eye/Face Washes
Catalog No. 19003209 Shop All Speakman Company Products
Click to view available options
Includes:
Flip-top Dust Covers
Activation:
Push Handle
Catalog No. Includes Activation
19-003-209 Flip-top Dust Covers Push Handle
1 options
Catalog No. 19-003-209 Supplier Speakman™ Supplier No. SE582
Includes
Emergency sign
Wall-mounted emergency eyewash
  • Stainless steel bowl
  • Dual aerated spray outlets
  • Wall mount comes with bracket and chrome plated tailpiece
  • Comfortably flush eyes for the ANSI recommended 15 minutes
Specifications
Activation Push Handle
Certifications/Compliance ANSI Z358.1
Flow Rate 3.6 gpm
Includes Flip-top Dust Covers
Mounting Wall
Pressure 30 psi
Product Type Eyewash