Learn More
Speakman™ SE-921 Counter Top Mounted Drench Hose
Catalog No. 19058328 Shop All Speakman Company Products
Click to view available options
Mounting:
Deck
Catalog No. Mounting
19-058-328 Deck
1 options
Catalog No. 19-058-328 Supplier Speakman™ Supplier No. SE921
Specifications
Certifications/Compliance ANSI/ISEA Z358.1 Certified
Flow Rate 1.6 gpm
Includes Singular, aerated spray head; Stay-open squeeze valve activation; 6-foot hose
Mounting Deck
Pressure 30 psi
Product Type Drench Hose