Learn More
Mitegen Spearlab Large Vessel Dewar
SDP
Catalog No. 501492137
Encompass_Preferred
Dimensions of the cylindrical cavity in this vessel are 6.3 inches in diameter by 3.8 inches deep, so that it easily holds 1400 ml of liquid nitrogen. Each dewar comes with a matching foam lid to insulate the contents from ambient air.
Catalog No. 50-149-2137 Supplier Mitegen Supplier No. 1020432
Dimensions of the cylindrical cavity in this vessel are 6.3 inches in diameter by 3.8 inches deep, so that it easily holds 1400 ml of liquid nitrogen. Each dewar comes with a matching foam lid to insulate the contents from ambient air.