Learn More
Mitegen Spearlab Standard Vessel Dewar
SDP
Catalog No. 501492136
Encompass_Preferred
This vessel, which holds 800 ml of liquid nitrogen. It replaces a large sized low profile glass dewar. The foam dewar has superior durability, safety, and thermal insulating properties. The standard vessel shape is circular, with a protruding handle. Each dewar comes with a matching foam lid to insulate the contents from ambient air. The cylindrical cavity in this vessel is 5.8 inches in diameter by 2.8 inches deep.
Catalog No. 50-149-2136 Supplier Mitegen Supplier No. 1020431
This vessel, which holds 800 ml of liquid nitrogen. It replaces a large sized low profile glass dewar. The foam dewar has superior durability, safety, and thermal insulating properties. The standard vessel shape is circular, with a protruding handle. Each dewar comes with a matching foam lid to insulate the contents from ambient air. The cylindrical cavity in this vessel is 5.8 inches in diameter by 2.8 inches deep.