Learn More
Medchemexpress LLC SPEB E.coli His-SU 50ug
SDP
Catalog No. 5000195945
Encompass_Preferred
SPEB is a recombinant E. coli agmatine ureohydrolase supplied as a His-SUMO-tagged protein for laboratory research. It catalyzes the conversion of agmatine to putrescine, a key step in polyamine biosynthesis, and is provided lyophilized for convenient storage and handling.
  • His-SUMO-tagged recombinant protein for affinity purification.
  • Expressed in an E. coli expression system to enable bacterial expression yields.
  • Catalyzes conversion of agmatine to putrescine in polyamine biosynthesis.
  • Lyophilized from PBS with 6% trehalose to enhance stability.
  • Supplied in a 50 μg research-scale quantity.
  • Stable at -20°C for up to 2 years; aliquot and freeze at -20°C or -80°C after reconstitution.
  • Intended for research use only.
Catalog No. 50-001-95945 Supplier Medchemexpress LLC Supplier No. HYP7207250UG
SPEB is a recombinant E. coli agmatine ureohydrolase supplied as a His-SUMO-tagged protein for laboratory research. It catalyzes the conversion of agmatine to putrescine, a key step in polyamine biosynthesis, and is provided lyophilized for convenient storage and handling.
  • His-SUMO-tagged recombinant protein for affinity purification.
  • Expressed in an E. coli expression system to enable bacterial expression yields.
  • Catalyzes conversion of agmatine to putrescine in polyamine biosynthesis.
  • Lyophilized from PBS with 6% trehalose to enhance stability.
  • Supplied in a 50 μg research-scale quantity.
  • Stable at -20°C for up to 2 years; aliquot and freeze at -20°C or -80°C after reconstitution.
  • Intended for research use only.