Learn More
ACROBiosystems SpeB  is a unique recombinantly produced cysteine protease that digests antibodies from many species and subclasses in the hinge region, producing a homogenous pool of Fab fragments. The enzyme requires reducing conditions for activity on I
Catalog No. 502253099
Encompass_Preferred
SpeB is derived from Streptococcus pyogenes. It is an important streptococcal virulence factor which cleaves human fibronectin and degrades vitronectin. It also cleaves human IL1B precursor to form biologically active IL1B and can induce apoptosis in human monocytes and epithelial cells in vitro, reduces phagocytic activity in monocytic cells. Thus, it may play a role in bacterial colonization, invasion, and inhibition of wound healing.
This item is not returnable. View return policy
Catalog No. 50-225-3099 Supplier ACROBiosystems Supplier No. SPBS51152000U
SpeB is derived from Streptococcus pyogenes. It is an important streptococcal virulence factor which cleaves human fibronectin and degrades vitronectin. It also cleaves human IL1B precursor to form biologically active IL1B and can induce apoptosis in human monocytes and epithelial cells in vitro, reduces phagocytic activity in monocytic cells. Thus, it may play a role in bacterial colonization, invasion, and inhibition of wound healing.