Learn More
ORS Nasco SPEC ASTR C/BL DUOFLX XTR 10EA
Catalog No. 502551588
Encompass_Preferred
Honeywell part# S1359C. Honeywell Uvex Astrospec 3000 Safety Glasses with Duoflex Temples Frame Color: Black Lens Type: Anti-fog Lens Tint: Clear Lens Coating: Uvextreme Anti-fog Certifications/Compliance: ANSI Z87+ and CSA Z94.3 standards Recommended Applications: Safety Temple Style: Duoflex
Catalog No. 50-255-1588 Supplier ORS Nasco Supplier No. 763S1359C
Add to Cart
Add to Cart
Honeywell part# S1359C. Honeywell Uvex Astrospec 3000 Safety Glasses with Duoflex Temples Frame Color: Black Lens Type: Anti-fog Lens Tint: Clear Lens Coating: Uvextreme Anti-fog Certifications/Compliance: ANSI Z87+ and CSA Z94.3 standards Recommended Applications: Safety Temple Style: Duoflex