Learn More
ORS Nasco SPEC ASTROSPEC OTG BLK/CLR1PR
Catalog No. 502551587
Encompass_Preferred
Honeywell part# S2500. Honeywell Safety Products Uvex Astro OTG 3001 Safety Glasses Black frame; Clear ultradura lens Frame Color: Black Lens Type: Scratch-resistant Lens Tint: Clear Frame Material: Nylon Lens Coating: Ultra-dura Hardcoat Lens Material: Polycarbonate UV Protection: 99.90%
Catalog No. 50-255-1587 Supplier ORS Nasco Supplier No. 763S2500
Honeywell part# S2500. Honeywell Safety Products Uvex Astro OTG 3001 Safety Glasses Black frame; Clear ultradura lens Frame Color: Black Lens Type: Scratch-resistant Lens Tint: Clear Frame Material: Nylon Lens Coating: Ultra-dura Hardcoat Lens Material: Polycarbonate UV Protection: 99.90%