Learn More
Starna Cells Inc SPEC CELL GLASS 100MM 10CM
SDP
Catalog No. NC9631455
Encompass_Preferred
Starna Standard Rectangular Cells, Window material Optical Glass, Usable range: 334 to 2500nm, Path Length: 100mm, External W and L and H, 12.5mm and 102.5mm and 45mm, Internal W and L, 10mm and 100mm, Nominal Volume: 35.00mL
Catalog No. NC9631455 Supplier Starna Cells Inc Supplier No. 1G100
Starna Standard Rectangular Cells, Window material Optical Glass, Usable range: 334 to 2500nm, Path Length: 100mm, External W and L and H, 12.5mm and 102.5mm and 45mm, Internal W and L, 10mm and 100mm, Nominal Volume: 35.00mL