Learn More
Hach Company SpecCheck Secondary Gel Standards Set, DPD Chlorine - LR
Supplier:  Hach Company 2635300
Encompass
DPD Chlorine, Low Range, 0-2.00 mg/L as CI₂. Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations. *This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6400