Learn More
Hach Company SpecCheck Gel Secondary Standard Set, Fluoride
Supplier:  Hach Company 2712500
Encompass
For Fluoride SPADNS method, 0-2.0 mg/L as F⁻ Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Catalog No. 50-199-6589