Learn More
Hach Company SpecCheck Secondary Gel Standards Set for Monochloramine & Free Ammonia.
Supplier:  Hach Company 2507500
Encompass
Monochloramine/Free Ammonia, 0-4.50 mg/L as Cl₂; 0-0.50 mg/L as NH₃-N. Secondary standards, used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6113