Learn More
Hach Company SpecCheck Gel Secondary Standard Set, Ozone, Mid-Range
Supplier:  Hach Company 2708000
Encompass
For Ozone Indigo method, MR, 0-.75 mg/L as O₃ Used to confirm consistent instrument response. Set contains four vials filled with gels that simulate the test color at various concentrations.
Catalog No. 50-199-6587