Learn More
Cole-Parmer Jenway 720 601 UV/Visible Diode Array Scanning Spectrophotometer
Catalog No. 501961214 Shop All Cole-Parmer Products
Encompass_Preferred
The multi-element detectors on these scanning spectrophotometers measures the entire wavelength range at once, reducing wait times. Plus, each dataset contains readings from the complete spectrum, making the data readily available for future analysis. The detector's orientation shields the array from stray light, allowing you to perform measurements with the lid open, quickly exchange samples, and decreases user errors in results. Small footprint takes up less bench space, the large color touch screen is easy to read, and the graphical interface is intuitive. The USB port on the rear connects with a printer (order separately), while the USB on the front accepts flash drives for data storage, quantitation curve uploads, and firmware updates. Measurement modes include photometrics, concentration, optical density (O.D. at 600 nm), quantitation, spectrum scanning, and kinetics.
Catalog No. 50-196-1214 Supplier Cole-Parmer Supplier No. 8305604
The multi-element detectors on these scanning spectrophotometers measures the entire wavelength range at once, reducing wait times. Plus, each dataset contains readings from the complete spectrum, making the data readily available for future analysis. The detector's orientation shields the array from stray light, allowing you to perform measurements with the lid open, quickly exchange samples, and decreases user errors in results. Small footprint takes up less bench space, the large color touch screen is easy to read, and the graphical interface is intuitive. The USB port on the rear connects with a printer (order separately), while the USB on the front accepts flash drives for data storage, quantitation curve uploads, and firmware updates. Measurement modes include photometrics, concentration, optical density (O.D. at 600 nm), quantitation, spectrum scanning, and kinetics.