Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wells Lamont™ Spec-Tec™ Sterile Cut Resistant Glove Liners - ANSI A2
GSA_VA

Catalog No. 19169810D Shop All Wells Lamont Industry Group Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-810D X-Large
19-169-810A Small
19-169-810B Medium
19-169-810C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-169-810D Supplier Wells Lamont™ Supplier No. M104XL
  • $1,021.00
    $761.65 / Pack of 20

    Save $259.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $3,550.00
    $2,649.65 / Case of 4 PK

    Save $900.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

HPPE fibers are comfortable and stretchy, while also providing ANSI Cut Level A2 protection. Each pair of Spec-Tec™ Sterile Cut Resistant Glove Liners is individually wrapped around tissue paper in a secure, sterile package.

Lightweight and comfortable

  • Sterile liners are constructed from a blend of Lycra™ and HPPE fibers
  • Liners provide ANSI Cut Level A2 protection
  • Individually wrapped around tissue paper in secure, sterile packaging
  • Single use in sterile environments
  • Launderable for reuse in non-sterile environments
  • Latex-free
  • Made in the USA

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Coating Material Uncoated
Color White
Cut Resistance Rating ANSI A2
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material HPPE, Lycra
Product Type Cut-resistant Glove Liners
Size X-Large
Sterility Sterile
For Use With (Application) Medical, Organ Tissue Processing, and Pharmaceuticals
Glove Cuff Style Knit
Hem Color Red
Inspected for Medical Use Yes
Packaging Type Individually Packed
Series M104
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.