Learn More
Sigma Aldrich Fine Chemicals Biosciences SpecacR Atlas 15T manual h1EA
Catalog No. 502908585
Encompass
Specac(R) Atlas 15T manual hydraulic press may be used for the fabrication of free-standing bio-composites by employing two bio-polyesters with different elastic moduli and reinforced with different graphene nanoplatelets. It may also be used to compress iron oxide granules to iron oxide discs to calculate the contact angle for the adsorption of heavy metals from wastewater using granulated iron oxide.
Catalog No. 50-290-8585 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8031031EA
Add to Cart
Add to Cart
Request a Quote
Specac(R) Atlas 15T manual hydraulic press may be used for the fabrication of free-standing bio-composites by employing two bio-polyesters with different elastic moduli and reinforced with different graphene nanoplatelets. It may also be used to compress iron oxide granules to iron oxide discs to calculate the contact angle for the adsorption of heavy metals from wastewater using granulated iron oxide.