Learn More
Sigma Aldrich Fine Chemicals Biosciences SpecacR mini pellet press1EA
Catalog No. 502908602
Encompass
The Specac Mini-Pellet Press is an entry-level laboratory hydraulic press for the preparation of high-quality 7mm KBr pellets for transmission FTIR analysis. With a rugged and durable design this press is intended for regular laboratory use with maintenance-fee operation.Specacs Mini-Pellet Press is designed to be used on a laboratory bench top but is small in size and light enough in weight to allow it to be moved to different points of use or stored when not required. Full hydraulic operation and an innovative sealed hydraulic unit allows the user to apply the required load for sample preparation through the simple rotation of a load knob. Reproducible load for pressing high-quality KBr pellets suitable for quantitative studies is facilitated by use of integrated pressure gauge. This pack includes the Mini-Pellet Press a 7mm Pellet Die Assembly (Z802557) 7mm Disc Holder for a standard 3 by 2 mount (Z802670) a pair of spare Pellet Ring Holders (Z802786) a Pestle and Mortar an
Catalog No. 50-290-8602 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8024411EA
The Specac Mini-Pellet Press is an entry-level laboratory hydraulic press for the preparation of high-quality 7mm KBr pellets for transmission FTIR analysis. With a rugged and durable design this press is intended for regular laboratory use with maintenance-fee operation.Specacs Mini-Pellet Press is designed to be used on a laboratory bench top but is small in size and light enough in weight to allow it to be moved to different points of use or stored when not required. Full hydraulic operation and an innovative sealed hydraulic unit allows the user to apply the required load for sample preparation through the simple rotation of a load knob. Reproducible load for pressing high-quality KBr pellets suitable for quantitative studies is facilitated by use of integrated pressure gauge. This pack includes the Mini-Pellet Press a 7mm Pellet Die Assembly (Z802557) 7mm Disc Holder for a standard 3 by 2 mount (Z802670) a pair of spare Pellet Ring Holders (Z802786) a Pestle and Mortar an