Learn More
Sigma Aldrich Fine Chemicals Biosciences SpecacR Omni Cell demounta1EA
Catalog No. 502906147
Encompass
Specacs demountable Omni Cell System presents a novel approach for the analysis of liquid mulls pastes and cast film samples in UV Vis NIR and IR transmission spectroscopy with one cell mount being suitable for all applications.This transmission cell is specially designed to be suitable with both fTIR and dispersive instruments and is available with a wide variety of high-quality window materials to meet your analytical needs. Includes front and back plates cell nest 4 quick release nuts bonded front PTfE gasket rear neoprene gasket and 2 PTfE Luer plugs. Please order the preferred windows and spacers separately.
Catalog No. 50-290-6147 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8037581EA
Specacs demountable Omni Cell System presents a novel approach for the analysis of liquid mulls pastes and cast film samples in UV Vis NIR and IR transmission spectroscopy with one cell mount being suitable for all applications.This transmission cell is specially designed to be suitable with both fTIR and dispersive instruments and is available with a wide variety of high-quality window materials to meet your analytical needs. Includes front and back plates cell nest 4 quick release nuts bonded front PTfE gasket rear neoprene gasket and 2 PTfE Luer plugs. Please order the preferred windows and spacers separately.