Learn More
Sigma Aldrich Fine Chemicals Biosciences SpecacR Omni Cell windows
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z8040882EA
Encompass
Specac(R)AOmni Cell windows [CaF2 41 mm x 23 mm x 4 mm] are accessories mainly for use with Specac(R) Omni Cell demountable cell system for IR spectroscopy analysis of liquids mulls pastes and cast film samples.
Catalog No. 50-290-8898