Learn More
Sigma Aldrich Fine Chemicals Biosciences SpecacR Omni Cell windows
Catalog No. 502891545
Encompass
Specac(R)AOmni Cell windows [KBr, 41 mm x 23 mm x 4 mm] are accessories mainly for use with Specac(R) Omni Cell demountable cell system for IR spectroscopy analysis of liquids, mulls, pastes, and cast film samples.
Catalog No. 50-289-1545 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z8039792EA
Specac(R)AOmni Cell windows [KBr, 41 mm x 23 mm x 4 mm] are accessories mainly for use with Specac(R) Omni Cell demountable cell system for IR spectroscopy analysis of liquids, mulls, pastes, and cast film samples.