Learn More
Analytical Sales & Services Special 2 Piece Ferrules for 1/8" OD Tubing
Catalog No. 5000805787
Encompass
Special 2 Piece Ferrules for 1/8" OD Tubing - to be used with Low Pressure Valves, VXL family
Catalog No. 50-008-05787 Supplier Analytical Sales & Services Supplier No. 77825
Special 2 Piece Ferrules for 1/8" OD Tubing - to be used with Low Pressure Valves, VXL family