Learn More
Hach Company Special Analog Insert.Mnt. ORP
Catalog No. 5000207629 Shop All Hach Company Products
Encompass
Hach Online Process ORP Sensor - Speciality Analog Insertion Mount ORP SensorSpecialty Online Process ORP Sensor - Gold ORP Electrode PEEK Housing Insertion Mount 4 5m Cable Analog Cable Connection
Catalog No. 50-002-07629 Supplier Hach Company Supplier No. RD2P6
Hach Online Process ORP Sensor - Speciality Analog Insertion Mount ORP SensorSpecialty Online Process ORP Sensor - Gold ORP Electrode PEEK Housing Insertion Mount 4 5m Cable Analog Cable Connection