Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Special Industrial Solvent SIS A-2 190 Proof, Greenfield

Catalog No. 502946011
Encompass
Change view
Click to view available options
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6011 Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6011 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P1250A2190DM55M

Please call customer service for current pricing. 1-800-766-7000

Specifications

Quantity 55 Gallon
Packaging Metal Drum
Boiling Point 176 °F (80 °C)
Percent Purity 190 Proof
Chemical Name or Material Special Industrial Solvent (SIS) Formula 902, 190 Proof
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.