Learn More
Perkin Elmer US LLC Special Optical Glass Macro Cuvette with PTFE Cover - 10 mm, Qty. 2
Catalog No. 502093276
Encompass_Preferred
These durable, fire-fused cells are standard spectrophotometer cuvettes with two optically-polished windows. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size Optical Glass 10 mm 12.5 mm 12.5mm 3.5 mL Macro
Catalog No. 50-209-3276 Supplier Perkin Elmer US LLC Supplier No. N4103010
These durable, fire-fused cells are standard spectrophotometer cuvettes with two optically-polished windows. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size Optical Glass 10 mm 12.5 mm 12.5mm 3.5 mL Macro