Learn More
Perkin Elmer US LLC Special Quartz Macro Cuvette with PTFE Cover - 100 mm, Qty. 1
Catalog No. 502093281
Encompass_Preferred
These durable, fire-fused cells are standard rectangular spectrophotometer cuvettes with two optically-polished windows. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size UV Quartz 100 mm 102.5 mm 12.5 mm 35.0 mL Macro
Catalog No. 50-209-3281 Supplier Perkin Elmer US LLC Supplier No. N4103015
These durable, fire-fused cells are standard rectangular spectrophotometer cuvettes with two optically-polished windows. They are made from the highest quality quartz and glass material available to ensure excellent transmission values and highly accurate results. Features of PerkinElmer Cuvettes: Material Path Length Outer Length Outer Width Capacity Size UV Quartz 100 mm 102.5 mm 12.5 mm 35.0 mL Macro