Learn More
Fritsch Milling And Sizing Inc Special Sealing System for Ball-Mills (Inert Gas)
SDP
Catalog No. 501534719
Encompass
Additional lock-system for all grinding bowls 500 ml, 250 ml, 80 ml volume (for the transport of the closed grinding bowl)
With this special Additional Lock-System, you can gas-tight seal your grinding bowls for transport between filling in the glove box and the mill. With an additional adapter, it can also be used for small grinding bowls.
Catalog No. 50-153-4719 Supplier Fritsch Milling And Sizing Inc Supplier No. 90.1400.00
Additional lock-system for all grinding bowls 500 ml, 250 ml, 80 ml volume (for the transport of the closed grinding bowl)
With this special Additional Lock-System, you can gas-tight seal your grinding bowls for transport between filling in the glove box and the mill. With an additional adapter, it can also be used for small grinding bowls.