Learn More
Electron Microscopy Sciences Embra Special Shape Grid EA1500, Copper
SDP
Catalog No. 5028776
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Size: 1500 µm
Catalog No. 50-287-76 Supplier Electron Microscopy Sciences Supplier No. EA1500CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Size: 1500 µm