Learn More
Electron Microscopy Sciences Embra Special Shape Grid Z600, Copper
SDP
Catalog No. 5028775
Encompass_Preferred
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Size: inner:600 µm, outer 900 µm, width 150 µm
Catalog No. 50-287-75 Supplier Electron Microscopy Sciences Supplier No. Z600CU
Veco Grids offer superior handling characteristics. Plus, with a 0.8 mil thickness, Veco Grids are the most rigid grids available. 3.05 mm diameter.
Material: Copper
Size: inner:600 µm, outer 900 µm, width 150 µm