Learn More
Mopec™ Special Stains Control Slides, Amyloid or Congo Red
SDP
Catalog No. 22457579 Shop All Mopec Products
Change view
Click to view available options
Quantity:
10 Slides/Cs.
100 Slides/Cs.
20 Slides/Cs.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-457-579 100 Slides/Cs.
22-457-504 10 Slides/Cs.
22-457-545 20 Slides/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-457-579 Supplier Mopec™ Supplier No. SC802100
Add to Cart
Add to Cart
This item is not returnable. View return policy
All positive control slides are on IHC Enhanced Hydrophilic Adhesive Slides using Formalin Fixed Paraffin Embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60°F to 80°F in a humidity controlled and light-deprived environment
  • Case of 10 contains 10 unstained slides and 1 stained slide
  • Case of 20 contains 20 unstained slides and 2 stained slides
  • Case of 100 contains 98 unstained slides and 2 stained slides
Specifications
Quantity 100 Slides/Cs.
Product Type Control Slide