Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Specially Denatured Alcohol 3A, 87.9 to 88.4%, MilliporeSigma™

Catalog No. MEX027220 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MEX027220 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MEX027220 Supplier MilliporeSigma Supplier No. EX027220

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Specially Denatured Alcohol 3A
Quantity 200 L
Physical Form Liquid
Assay Percent Range 87.9 to 88.4%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.