Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Specially Denatured Alcohol SDA 3A 190 Proof, Greenfield

Catalog No. 502946001
Encompass
Change view
Click to view available options
Metal Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-294-6001 Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-294-6001 Supplier Thermo Scientific Production Chemicals and Services Supplier No. P12303A190DM55M

Please call customer service for current pricing. 1-800-766-7000

missing translation for 'validMainframeMsg'

Specifications

Quantity 55 Gallon
Packaging Metal Drum
Boiling Point 173.12 °F (78.4 °C)
Percent Purity 190 Proof
Chemical Name or Material Special Denatured Alcohol (SDA) 3A, 190 Proof
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.