Learn More
Bachem H-Glu(bNA)-OH
SDP
Catalog No. 501947046
Encompass_Preferred
H-Glu(bNA)-OH, 14525-44-1, C15H16N2O3, Mr 272.30, 1g. Specific and sensitive substrate for the determination of serum g-glutamyl transferase activity.
Catalog No. 50-194-7046 Supplier Bachem Supplier No. 4003731.1000
H-Glu(bNA)-OH, 14525-44-1, C15H16N2O3, Mr 272.30, 1g. Specific and sensitive substrate for the determination of serum g-glutamyl transferase activity.