Learn More
Cole-Parmer Gardco WG-SS-8.32 Specific Gravity Cup, 8.32 mL, 1.2% Tolerance; U.S. use
Catalog No. NC0397349 Shop All Cole-Parmer Products
Encompass_Preferred
Use these precision-made stainless steel cups with your laboratory balance to determine specific gravity or pounds per gallon. Simply tare balance to cup weight, then weigh cup filled with sample. Order cups for U.S. Standard or British Standard determinations. All cups meet MIL STD 45662A and are NIST-traceable.
Catalog No. NC0397349 Supplier Cole-Parmer Supplier No. 3800000
Use these precision-made stainless steel cups with your laboratory balance to determine specific gravity or pounds per gallon. Simply tare balance to cup weight, then weigh cup filled with sample. Order cups for U.S. Standard or British Standard determinations. All cups meet MIL STD 45662A and are NIST-traceable.