Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Specific Gravity Standard, 1.20 at 60°F/60°F, Reagents

Catalog No. 01333483
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-333-483 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-483 Supplier Reagents Holdings Llc Supplier No. CS612060500A
Add to Cart
Add to Cart

Specific Gravity Standard, 1.20 at 60°F/60°F, Reagent, use for Specific gravity calibration. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Specific Gravity Standard, 1.20 at 60°F/60°F
Packaging Poly Bottle
CAS 7732-18-5, 7647-15-6
Specific Gravity 1.2
Physical Form Liquid
Color Colorless
pH 6.5 to 8
Boiling Point 103°C
Solubility Completely miscible
Quantity 500 mL
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.