Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Specific Gravity Standard, Sp. Gr. = 1.10 at 60°F/60°F, Ricca Chemical
SDP

Catalog No. 799211032 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
7992110-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 7992110-32 Supplier Ricca Chemical Company Supplier No. 7992.11032
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 81.0, 16.54
CAS Max % 85.0, 17.22
Physical Form Liquid
Color Colorless
pH > 12
Melting Point Approximately 0°C
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Analytical Standard
Quantity 1 L
Grade Laboratory
Chemical Name or Material Specific Gravity Standard, Sp. Gr. = 1.10 at 60°F/60°F
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.