Learn More
Perkin Elmer US LLC Indicating Glass Oxygen/Moisture/Hydrocarbon Triple Trap, Helium-Specific
Catalog No. 509048030
Encompass_Preferred
The glass indicating trap clearly shows when the filter needs to be replaced with the use of color changes. The packing material is a silica-based environmentally friendly substitute for cobalt dioxide (blue) in the moisture indicator. Available as a kit with the necessary 1/8 brass connectors, and as a replacement trap, this system is easy to install.
Catalog No. 50-904-8030 Supplier Perkin Elmer US LLC Supplier No. N9306107
The glass indicating trap clearly shows when the filter needs to be replaced with the use of color changes. The packing material is a silica-based environmentally friendly substitute for cobalt dioxide (blue) in the moisture indicator. Available as a kit with the necessary 1/8 brass connectors, and as a replacement trap, this system is easy to install.