Learn More
Electron Microscopy Sciences Specimen Clean Baskets
SDP
Supplier:  Electron Microscopy Sciences 7019026
Encompass
This basket is brass with a stainless steel mesh attached to both ends. To use, simply screw both halves together and you can keep small specimens for long periods of time during the process of fixing, dehydration or even critical point drying. It can also be used to hold small parts for ultrasonic cleaning. Mesh opening: 300 micron. Measurements: Inner Diameter 12.5 mm, Outer Diameter 16 mm, Height 16 mm.
Catalog No. 50-287-98