Learn More
McKesson General Medical McKesson Commode Specimen Collector, 800 mL Capacity, Polypropylene Material, 100 Per Case
Catalog No. NC1553544
Encompass_Preferred
The McKesson Commode Specimen Collector is a non-sterile, polypropylene device designed for urine and stool sample collection. With a graduated capacity of 800 mL (27 oz.), it features clear markings in ounces and cubic centimeters for precise measurement. Its single-patient use design and compatibility with standard commodes make it an essential tool for both clinical and home care settings. The collector is also latex-free, minimizing allergy risks, and comes packaged with 100 units per case for convenience.
Catalog No. NC1553544 Supplier McKesson General Medical Supplier No. 487960
The McKesson Commode Specimen Collector is a non-sterile, polypropylene device designed for urine and stool sample collection. With a graduated capacity of 800 mL (27 oz.), it features clear markings in ounces and cubic centimeters for precise measurement. Its single-patient use design and compatibility with standard commodes make it an essential tool for both clinical and home care settings. The collector is also latex-free, minimizing allergy risks, and comes packaged with 100 units per case for convenience.