Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Specimen Container, Graduated
SDP

Catalog No. 22024782
Change view
Click to view available options
Capacity (Metric):
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-024-782 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-024-782 Supplier StatLab Supplier No. CT6200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Graduated plastic specimen container with tamper-proof identification label.

  • Sterile 120 mL (4 oz.) plastic specimen container for secure specimen collection
  • Features graduated markings for convenient volume measurement
  • Includes a tamper-proof sticker with patient identification fields
  • Packaged with 300 containers/Cs. for laboratory and clinical use

Specifications

Product Type Specimen Container Vial
Closure Included Yes
Graduated Yes
Material Polypropylene
Shape Cylindrical
Sterility Sterile
Capacity (Metric) 120 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.