Learn More
Alkali Scientific ASI™ Specimen Cup (Container), DuoClick, Leak-Resistant | Pathology, Microbiology, Histology, Sterile, 90mL x 48mm, 400/Case, Blue
SDP
Catalog No. 503028224
Encompass_Preferred
Alkali Scientific™ Specimen Containers are specifically designed for collection and trasnport of Microbiology, Urinalysis, Pathology, Drug Testing and Histology specimens and are leak proof.  All containers are available in Sterile and Non-Sterile formats. Sterility Certification for each lot is provided upon request. Leak Resistant - 95 Kpa compliant Exceptional clarity Proof of closure Graduation marking (3 sides) Top and bottom grip-knurl Manufactured from pure virgin medical grade plastic Quality Design, Dependable Service, Reasonably Priced
Catalog No. 50-302-8224 Supplier Alkali Scientific Supplier No. SCDC9048BS
Alkali Scientific™ Specimen Containers are specifically designed for collection and trasnport of Microbiology, Urinalysis, Pathology, Drug Testing and Histology specimens and are leak proof.  All containers are available in Sterile and Non-Sterile formats. Sterility Certification for each lot is provided upon request. Leak Resistant - 95 Kpa compliant Exceptional clarity Proof of closure Graduation marking (3 sides) Top and bottom grip-knurl Manufactured from pure virgin medical grade plastic Quality Design, Dependable Service, Reasonably Priced